anti-IFN gamma, FITC, Clone: XMG1.2, EMD Millipore™
Rat Monoclonal Antibody
Manufacturer:  Millipore Sigma MABF1517
 View more versions of this product
Catalog No. MABF1517MI
Description
Description
Specifically detects IFN gamma Clone: XMG1.2 in Mouse samples, and it is validated for Flow Cytometry
The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells, which allow the cytokine to modulate innate immunity, when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein, e.g. by flow cytometry, as well as for quantitative analysis of the secreted protein by ELISA, when paired with an appropriate secondary antibody. This clone is also widely used for neutralization of the functional activity of IFN-g in a variety of assays.
Specifications
Specifications
IFN gamma
XMG1.2
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Purified
Rat
IgG1, κ
25μg
Inflammation, Immunology
Primary
Murine
Flow Cytometry
FITC
P01580
Ifng
Proprietary immunogen
Purified by size-exclusion chromatography
RUO
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Monoclonal