missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IFI202b, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 273-290 of the mouse Ifi202b protein.
Antigen IFI202b
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227066 Novus Biologicals Canada
NBP227066
0.1mg Each for $309.05
NBP227066SS Novus Biologicals Canada
NBP227066SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
IFI202b
Purified
RUO
26388
0.5mg/mL
A synthetic peptide corresponding to amino acids 273-290 of the mouse Ifi202b protein.
Polyclonal
Primary
Western Blot
100μg in 200μL PBS containing 0.05% BSA.