Please login to your online account to display your discounted pricing

anti-IDH2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222166SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222166SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. P48735
Gene ID (Entrez) 3418
Monoclonal or Polyclonal Polyclonal
Test Specificity Human, mouse and rat. Predicted to react with monkey, hamster, Guinea pig and rabbit based on 100% sequence homology.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Concentration 1.0mg/mL
Conjugate Unlabeled
Quantity 0.025mL
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human IDH2 protein (between residues 375-452) [UniProt P48735].
Format Affinity Purified
Antigen IDH2
Regulatory Status RUO