Please login to your online account to display your discounted pricing

anti-IDH2, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human IDH2 protein (between residues 375-452) [UniProt P48735].
Antigen IDH2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222166

 
novus biologicals canada
NBP222166
Unlabeled 0.1mL Each for $309.05

NBP222166SS

 
novus biologicals canada
NBP222166SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human IDH2 protein (between residues 375-452) [UniProt P48735].
Antigen IDH2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human, mouse and rat. Predicted to react with monkey, hamster, Guinea pig and rabbit based on 100% sequence homology.
Gene Accession No. P48735
Gene ID (Entrez) 3418
Formulation PBS
Concentration 1.0mg/mL