Please login to your online account to display your discounted pricing

anti-ID2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 60-110 of human ID2 was used as the immunogen for this antibody.
Antigen ID2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227194

 
novus biologicals canada
NBP227194
Unlabeled 0.1mg Each for $257.41

NBP227194SS

 
novus biologicals canada
NBP227194SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 60-110 of human ID2 was used as the immunogen for this antibody.
Antigen ID2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, orangutan, rhesus monkey, white-cheeked gibbon, macaque, golden hamster, and mouse, 94% in pig, panda, cow, horse, and rat, 89% in new world monkey, platypus, wild turkey, chicken and Japanese quail, 83% in zebra finch and opossum, and 61% homologous in Carolina anole.
Gene ID (Entrez) 3398
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL