Please login to your online account to display your discounted pricing

anti-ICK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223656

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223656

  • / Each
 

Description & Specifications

Specifications

Antigen ICK
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Host Species Rabbit
Immunogen A portion of amino acid 380-430 of human ICK protein was used as the immunogen.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 22858
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), monkey, mouse and dog, 83% homologous in chicken and xenopus, 72% homologous in pufferfish.
Target Species Bovine
Target Species Human
Target Species Primate