Please login to your online account to display your discounted pricing

anti-ICK, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 380-430 of human ICK protein was used as the immunogen.
Antigen ICK
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223656

 
novus biologicals canada
NBP223656
Unlabeled 0.1mL Each for $259.02

NBP223656SS

 
novus biologicals canada
NBP223656SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 380-430 of human ICK protein was used as the immunogen.
Antigen ICK
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), monkey, mouse and dog, 83% homologous in chicken and xenopus, 72% homologous in pufferfish.
Gene ID (Entrez) 22858
Formulation 100μL of crude rabbit sera with 50% glycerol