Please login to your online account to display your discounted pricing

anti-ICAM-1/CD54, Clone: BBIG-I1, R&D Systems

$320.68 - $320.68

Specifications

Antigen ICAM-1/CD54
Clone BBIG-I1
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BBA3

 
r & d systems
RDSBBA3
Unlabeled 100μg Each for $320.68
Description & Specifications

Specifications

Antigen ICAM-1/CD54
Clone BBIG-I1
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Activated HUVEC human umbilical vein endothelial cells.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Blocking (Adhesion), Immunocytochemistry, Immunoprecipitation, Western Blotting