Please login to your online account to display your discounted pricing

anti-Human ZAP70, Polyclonal, R&D Systems

$467.98 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P43403
Gene ID (Entrez) 7535
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF3709

 
r & d systems
RDSAF3709
Unlabeled 100μg Each for $467.98

AF6375

 
r & d systems
RDSAF6375
Unlabeled 100μg Each for $467.98
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P43403
Gene ID (Entrez) 7535

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human ZAP70 , Trp163-Cys254, Accession No. P43403

Applications:

Immunohistochemistry, Western Blotting