anti-Human VCAM-1/CD106, Polyclonal, R&D Systems
Manufacturer:  R & D Systems RDSAF809
 View more versions of this product
Catalog No. AF809
Specifications
Description
Antibody; Primary; anti-VCAM-1/CD106, Polyclonal, R&D Systems; 100μg; Unlabeled; Host: Sh; Isotype: IgG; Applications: Blck (Ad), WB; Species Reactivity: Hu
VCAM-1/CD106 Polyclonal antibody AF809 Specifically detects VCAM-1/CD106 in Human samples. It is validated for Blocking (Adhesion), Western Blotting
Description & Specifications
Specifications
VCAM-1/CD106
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Sheep
IgG
RUO
Polyclonal
Human
Unlabeled
Affinity Purified
Chinese hamster ovary cell line (CHO)-derived recombinant human VCAM-1, Extracellular domain
100μg
Primary
7412