missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human VCAM-1/CD106, Polyclonal, R&D Systems

$252.10 - $467.46

Specifications

Gene ID (Entrez) 7412
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
AF809 R & D Systems
RDSAF809
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $467.46
BBA19 R & D Systems
RDSBBA19
Lyophilized from a 0.2μm filtered solution in Serum. Each for $252.10
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:Chinese hamster ovary cell line CHO-derived recombinant human VCAM-1

Applications:

Immunohistochemistry, Western Blotting

Description & Specifications

Specifications

7412