missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human Survivin, Polyclonal, R&D Systems

$376.73 - $481.01

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 332
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
AF886 R & D Systems
RDSAF886
Unlabeled 100μg Each for $376.73
AF6471 R & D Systems
RDSAF6471
Unlabeled 100μg Each for $481.01
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human Survivin

Applications:

Flow Cytometry, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
332