Please login to your online account to display your discounted pricing

anti-Human SMAC/Diablo, Polyclonal, R&D Systems

$368.77 - $458.10

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9NR28
Gene ID (Entrez) 56616
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF789

 
r & d systems
RDSAF789
Unlabeled 100μg Each for $368.77

AF7891

 
r & d systems
RDSAF7891
Unlabeled 100μg Each for $458.10
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9NR28
Gene ID (Entrez) 56616

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human SMAC/Diablo, Ala56-Asp239, Accession No. Q9NR28

Applications:

Immunohistochemistry, Western Blotting