Please login to your online account to display your discounted pricing

anti-Human SMAC/Diablo, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF7891

 View more versions of this product

Catalog No. AF7891

  • / Each
 

Description & Specifications

Specifications

Antigen SMAC/Diablo
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9NR28
Host Species Goat
Immunogen E. coli-derived recombinant human SMAC/Diablo, Ala56-Asp239, Accession No. Q9NR28
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 56616
Target Species Human

SMAC/Diablo Polyclonal antibody AF7891 Specifically detects SMAC/Diablo in Human samples. It is validated for Immunohistochemistry, Western Blotting