missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human R-Spondin 3, Polyclonal, R&D Systems

$454.80 - $454.80

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9BXY4
Gene ID (Entrez) 84870
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
AF35001 R & D Systems
RDSAF35001
Each for $454.80
AF3500 R & D Systems
RDSAF3500
Each for $454.80
Specifications

Description

Host Species: Sheep
Species Reactivity:Human
Immunogen:Chinese hamster ovary cell line CHO-derived recombinant human R-Spondin 3, Gln22-His272, Accession No. Q9BXY4

Applications:

Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Q9BXY4
84870