Please login to your online account to display your discounted pricing

anti-Human R-Spondin 3, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF3500

 View more versions of this product

Catalog No. AF3500

  • / Each
 

Description & Specifications

Specifications

Antigen R-Spondin 3
Regulatory Status RUO
Cross Reactivity Human
Quantity 100μg
Format Affinity Purified
Host Species Goat
Isotype IgG
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Mouse myeloma cell line NS0-derived recombinant human&No.160;R-Spondin 3, Ser21-Gly209, Accession No. Q9BXY4
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9BXY4
Gene ID (Entrez) 84870
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.

R-Spondin 3 Polyclonal antibody AF3500 Specifically detects R-Spondin 3 in Human samples. It is validated for Western Blotting