Please login to your online account to display your discounted pricing

anti-Human PTP1B, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF13662

 View more versions of this product

Catalog No. AF13662

  • / Each
 

Description & Specifications

Specifications

Antigen PTP1B
Cross Reactivity Human
Regulatory Status RUO
Format Affinity Purified
Immunogen E. coli-derived recombinant human PTP1B , Glu2-Thr435, Accession No. P18031
Host Species Goat
Quantity 100μg
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 5770
Monoclonal or Polyclonal Polyclonal
Isotype IgG
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P18031

PTP1B Polyclonal antibody AF13662 Specifically detects PTP1B in Human samples. It is validated for Western Blotting