missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human PTP1B, Polyclonal, R&D Systems

$376.73 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 5770
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
AF1366 R & D Systems
RDSAF1366
Unlabeled 100μg Each for $376.73
AF13662 R & D Systems
RDSAF13662
Unlabeled 100μg Each for $467.98
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human PTP1B , Glu2-Thr435, Accession No. P18031

Applications:

Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
5770