Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat p70 S6 Kinase, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF8964

 View more versions of this product

Catalog No. AF8964

  • / Each
 

Description & Specifications

Specifications

Antigen p70 S6 Kinase
Format Affinity Purified
Host Species Goat
Isotype IgG
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. M60725
Immunogen E. coli-derived recombinant human p70&No.160;S6K, Accession No. M60725
Quantity 100μg
Primary or Secondary Primary
Gene ID (Entrez) 83840
Conjugate Unlabeled

p70 S6 Kinase Polyclonal antibody AF8964 Specifically detects p70 S6 Kinase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting