Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat p70 S6 Kinase, Polyclonal, R&D Systems

$376.73 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. M60725
Gene ID (Entrez) 83840
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF8964

 
r & d systems
RDSAF8964
Unlabeled 100μg Each for $467.98

AF8962

 
r & d systems
RDSAF8962
Unlabeled 100μg Each for $376.73
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. M60725
Gene ID (Entrez) 83840

Host Species: Goat
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human p70&No.160;S6K, Accession No. M60725

Applications:

Immunohistochemistry, Western Blotting