missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat p38 gamma, Polyclonal, R&D Systems

$366.12 - $454.80

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P53778
Gene ID (Entrez) 6300
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
AF1347 R & D Systems
RDSAF1347
Unlabeled 100μg Each for $366.12
AF1644 R & D Systems
RDSAF1644
Unlabeled 100μg Each for $454.80
Specifications

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human p38γ, Accession No. P53778

Applications:

Immunohistochemistry, Western Blotting, Simple Western

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P53778
6300