Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat p38 gamma, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF1644

 View more versions of this product

Catalog No. AF1644

  • / Each
 

Description & Specifications

Specifications

Antigen p38 gamma
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P53778
Host Species Goat
Immunogen E. coli-derived recombinant human p38γ, Met1-Leu367, Accession No. P53778
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6300
Target Species Human
Target Species Murine
Target Species Rat

p38 gamma Polyclonal antibody AF1644 Specifically detects p38 gamma in Human, Mouse, Rat samples. It is validated for Western Blotting