Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat MKK6, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF1604

 View more versions of this product

Catalog No. AF1604

  • / Each
 

Description & Specifications

Specifications

Antigen MKK6
Format Affinity Purified
Immunogen E. coli-derived recombinant human MKK6, Gly6-Asp334, Accession No. AAC50388
Isotype IgG
Quantity 50μg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. AAC50388
Host Species Rabbit
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 26399

MKK6 Polyclonal antibody AF1604 Specifically detects MKK6 in Human, Mouse, Rat samples. It is validated for Western Blotting