missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat MKK6, Polyclonal, R&D Systems

$376.73 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
AF16041 R & D Systems
RDSAF16041
P52564.1 5608 Each for $467.98
AF1604 R & D Systems
RDSAF1604
AAC50388 26399 Each for $376.73
Specifications

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human MKK6, Gly6-Asp334, Accession No. AAC50388

Applications:

Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.