Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat IKK gamma, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF2684

 View more versions of this product

Catalog No. AF2684

  • / Each
 

Description & Specifications

Specifications

Antigen IKK gamma
Format Affinity Purified
Host Species Goat
Immunogen E. coli-derived recombinant human IKKγ, Met1-Glu419, Accession No. Q9Y6K9
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9Y6K9
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8517

IKK gamma Polyclonal antibody AF2684 Specifically detects IKK gamma in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Simple Western, Western Blotting