missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat IKK gamma, Polyclonal, R&D Systems

$462.39 - $467.46

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
AF2684 R & D Systems
RDSAF2684
Q9Y6K9 8517 Each for $467.46
AF4365 R & D Systems
RDSAF4365
O88522 309295 Each for $462.39
Specifications

Description

Host Species: Goat
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human IKKγ, Met1-Glu419, Accession No. Q9Y6K9

Applications:

Immunocytochemistry, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.