Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat ERK2, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF1230

 View more versions of this product

Catalog No. AF1230

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Quantity 100μg
Host Species Rabbit
Immunogen E. coli-derived recombinant human ERK2, Met1-Ser360, Accession No. P28482
Format Affinity Purified
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 26413
Monoclonal or Polyclonal Polyclonal
Antigen ERK2
Regulatory Status RUO
Gene Accession No. P28482
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.

ERK2 Polyclonal antibody AF1230 Specifically detects ERK2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting