Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat ERK2, Polyclonal, R&D Systems

$368.77 - $458.10

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P28482
Gene ID (Entrez) 26413
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF12301

 
r & d systems
RDSAF12301
Unlabeled 100μg Each for $458.10

AF1230

 
r & d systems
RDSAF1230
Unlabeled 100μg Each for $368.77
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P28482
Gene ID (Entrez) 26413

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human ERK2, Met1-Ser360, Accession No. P28482

Applications:

Immunohistochemistry, Western Blotting