Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat ERK2, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF12301

 View more versions of this product

Catalog No. AF12301

  • / Each
 

Description & Specifications

Specifications

Immunogen E. coli-derived recombinant human ERK2, Met1-Ser360, Accession No. P28482
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen ERK2
Format Affinity Purified
Quantity 100μg
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 26413
Isotype IgG
Host Species Goat
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P28482
Monoclonal or Polyclonal Polyclonal

ERK2 Polyclonal antibody AF12301 Specifically detects ERK2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting