missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat Akt2, Polyclonal, R&D Systems

$376.73 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P31751
Gene ID (Entrez) 208
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF2315 R & D Systems
RDSAF2315
50μg Each for $376.73
AF23151 R & D Systems
RDSAF23151
100μg Each for $467.98
Specifications

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant recombinant human Akt2, Accession No. P31751

Applications:

Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P31751
208