Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat Akt1, Polyclonal, R&D Systems

$376.73 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P31749
Gene ID (Entrez) 25233
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF1775

 
r & d systems
RDSAF1775
Unlabeled 50μg Each for $376.73

AF17751

 
r & d systems
RDSAF17751
Unlabeled 100μg Each for $467.98
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P31749
Gene ID (Entrez) 25233

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human Akt1, Ser2-Ala480, Accession No. P31749

Applications:

Western Blotting