missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse/Rat AIF, Polyclonal, R&D Systems

$376.73 - $428.87

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. O95831
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gene ID (Entrez) Price Quantity  
Catalog Number Mfr. No. Gene ID (Entrez) Price Quantity  
AF1457 R & D Systems
RDSAF1457
83833 Each for $376.73
AF5824 R & D Systems
RDSAF5824
9131 Each for $428.87
Specifications

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:E. coli-derived recombinant human AIF, Glu121-Asp613, Accession No. O95831

Applications:

Immunocytochemistry, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
O95831