missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse Caspase-8, Polyclonal, R&D Systems

$376.73 - $481.01

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
Catalog Number Mfr. No. Gene Accession No. Gene ID (Entrez) Price Quantity  
AF1650 R & D Systems
RDSAF1650
Q14790 12370 Each for $376.73
AF705 R & D Systems
RDSAF705
AAC50645 841 Each for $481.01
Specifications

Description

Host Species: Goat
Species Reactivity:Human, Mouse
Immunogen:E. coli-derived recombinant human Caspase-8 , Ser234-Asp496, Accession No. AAC50645

Applications:

Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.