missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human/Mouse Bcl-w, Polyclonal, R&D Systems

$366.12 - $454.80

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gene ID (Entrez) Price Quantity  
Catalog Number Mfr. No. Gene ID (Entrez) Price Quantity  
AF824 R & D Systems
RDSAF824
12050 Each for $366.12
AF8241 R & D Systems
RDSAF8241
599 Each for $454.80
Specifications

Description

Host Species: Goat
Species Reactivity:Human, Mouse
Immunogen:E. coli-derived recombinant human Bcl-w, Ala2-Thr172, Accession No. Q92843

Applications:

Immunohistochemistry

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.