Please login to your online account to display your discounted pricing

anti-Human/Mouse Bcl-w, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF8241

 View more versions of this product

Catalog No. AF8241

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-w
Format Affinity Purified
Host Species Goat
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 599
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q92843
Immunogen E. coli-derived recombinant human Bcl-w, Ala2-Thr172, Accession No. Q92843

Bcl-w Polyclonal antibody AF8241 Specifically detects Bcl-w in Human, Mouse samples. It is validated for Immunohistochemistry