Please login to your online account to display your discounted pricing

anti-Human Mcl-1, Polyclonal, R&D Systems

$458.10 - $458.10

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 4170
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF8281

 
r & d systems
RDSAF8281
Unlabeled 100μg Each for $458.10
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 4170

Host Species: Rabbit
Species Reactivity:Human
Immunogen:KLH-coupled human Mcl-1 synthetic peptide, RQSLEIISRYLREQATGAKDC

Applications:

Western Blotting