missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human IL-4 R alpha, Polyclonal, R&D Systems

$467.98 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P24394
Gene ID (Entrez) 3566
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF6844 R & D Systems
RDSAF6844
100μg Each for $467.98
AB230NA R & D Systems
RDSAB230NA
1mg Each for $467.98
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL&No.x2011;4&No.160;Rα, Gly24-His232, Accession No. P24394

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P24394
3566