missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human IL-1 alpha/IL-1F1, Polyclonal, R&D Systems

$415.83 - $479.45

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q53QF9
Gene ID (Entrez) 3552
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB200NA R & D Systems
RDSAB200NA
1mg Each for $415.83
AF200NA R & D Systems
RDSAF200NA
100μg Each for $479.45
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human IL-1α/IL-1F1, Ser113-Ala271, Accession No. Q53QF9

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Q53QF9
3552