Please login to your online account to display your discounted pricing

anti-Human IL-1 alpha/IL-1F1, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF200NA

 View more versions of this product

Catalog No. AF200NA

  • / Each
 

Description & Specifications

Specifications

Immunogen E. coli-derived recombinant human IL-1α/IL-1F1, Ser113-Ala271, Accession No. Q53QF9
Isotype IgG
Host Species Goat
Quantity 100μg
Format Affinity Purified
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 3552
Monoclonal or Polyclonal Polyclonal
Antigen IL-1 alpha/IL-1F1
Regulatory Status RUO
Cross Reactivity Human
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q53QF9

IL-1 alpha/IL-1F1 Polyclonal antibody AF200SP Specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Immunocytochemistry, Neutralization, Western Blotting