missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human ICAM-3/CD50, Polyclonal, R&D Systems

$467.98 - $475.80

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3385
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
AF813 R & D Systems
RDSAF813
Unlabeled 100μg Each for $467.98
AF715 R & D Systems
RDSAF715
Unlabeled 100μg Each for $475.80
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:Mouse myeloma cell line NS0-derived recombinant human ICAMG–3/CD50, Met1-His485, Accession No. CAA49473

Applications:

Flow Cytometry, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
3385