missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Human ICAM-3/CD50, Biotin, Polyclonal, R&D Systems

$508.39 - $547.50

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3385
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
BAF715 R & D Systems
RDSBAF715
Biotin 50μg Each for $508.39
BAF813 R & D Systems
RDSBAF813
Biotin 50μg Each for $547.50
Specifications

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:Mouse myeloma cell line NS0-derived recombinant human ICAMG–3/CD50, Gln30-His485, Accession No. CAA49473

Applications:

ELISA Detection, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
3385