Please login to your online account to display your discounted pricing

anti-Human ICAM-3/CD50, Biotin, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSBAF813

 View more versions of this product

Catalog No. BAF813

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Cross Reactivity Human
Format Affinity Purified
Host Species Goat
Primary or Secondary Primary
Gene ID (Entrez) 3385
Conjugate Biotin
Antigen ICAM-3/CD50
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene Accession No. CAA49473
Immunogen Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50, Gln30-His485, Accession No. CAA49473
Quantity 50μg
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal