Please login to your online account to display your discounted pricing

anti-Human CD117/c-kit, Polyclonal, R&D Systems

$368.77 - $470.86

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P10721
Gene ID (Entrez) 3815
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AF3267

 
r & d systems
RDSAF3267
Unlabeled 50μg Each for $368.77

AF332

 
r & d systems
RDSAF332
Unlabeled 100μg Each for $470.86
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P10721
Gene ID (Entrez) 3815

Host Species: Rabbit
Species Reactivity:Human
Immunogen:Human SCF R synthetic peptide, Asp716-Pro726, Accession No. P10721

Applications:

Immunohistochemistry, Western Blotting