Please login to your online account to display your discounted pricing

anti-Human CD117/c-kit, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF332

 View more versions of this product

Catalog No. AF332

  • / Each
 

Description & Specifications

Specifications

Antigen CD117/c-kit
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P10721
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SCF R/c-kit, Gln26-Thr520, Accession No. P10721
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3815
Target Species Human

CD117/c-kit Polyclonal antibody AF332 Specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Simple Western, Western Blotting