Please login to your online account to display your discounted pricing

anti-HtrA3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 273-287 (IVSTAQREGRELGLR) of human HtrA3 were used as immunogen.
Antigen HtrA3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001151

 
novus biologicals canada
NB6001151
Unlabeled 0.1mg Each for $255.80

NB6001151SS

 
novus biologicals canada
NB6001151SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 273-287 (IVSTAQREGRELGLR) of human HtrA3 were used as immunogen.
Antigen HtrA3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity The amino acid sequence is 100% homologous in HtrA3, 80% homologous in HtrA1 and 71% homologous in HtrA2 and HtrA4. Cross-reacts with Human, Mouse and Rat. Predicted to react with Chicken and Cat.
Gene Accession No. P83110
Gene ID (Entrez) 94031
Formulation 100μg of antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL