Please login to your online account to display your discounted pricing

anti-HTRA1/PRSS11, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Gen Bank no NP_002766.1. This peptide sequence is in the serine protease domain of the HtrA1 protein.
Antigen HTRA1/PRSS11
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223869SS

 
novus biologicals canada
NBP223869SS
Unlabeled 0.025mL Each for $109.00

NBP223869

 
novus biologicals canada
NBP223869
Unlabeled 0.1mL Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Gen Bank no NP_002766.1. This peptide sequence is in the serine protease domain of the HtrA1 protein.
Antigen HTRA1/PRSS11
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Test Specificity This antibody is reactive to Chimpanzee, Cow, Dog, Human, Mouse and Rat.
Gene Accession No. Q92743
Gene ID (Entrez) 5654
Formulation 0.1mL; crude rabbit sera with 50% glycerol