Please login to your online account to display your discounted pricing

anti-HSP90 beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177561SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177561SS

  • / Each
 

Description & Specifications

Specifications

Antigen HSP90 beta
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.14mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P08238
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Hsp90B protein (between residues 1-50) [UniProt P08238]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3326
Target Species Human
Target Species Murine
Target Species Rat