missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HSP90 beta, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Hsp90B protein (between residues 650-724) [UniProt P08238]
Antigen HSP90 beta
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP177563 Novus Biologicals Canada
NBP177563
0.1mL Each for $309.05
NBP177563SS Novus Biologicals Canada
NBP177563SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
HSP90 beta
Affinity Purified
RUO
Human, mouse, rat and primate. Immunogen has 86% identity to cow.
3326
1mg/mL
A synthetic peptide made to a C-terminal portion of the human Hsp90B protein (between residues 650-724) [UniProt P08238]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
P08238
PBS, 30% glycerol