Please login to your online account to display your discounted pricing

anti-HSP90 beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177563SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177563SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Formulation PBS, 30% glycerol
Test Specificity Human, mouse, rat and primate. Immunogen has 86% identity to cow.
Gene ID (Entrez) 3326
Conjugate Unlabeled
Gene Accession No. P08238
Immunogen A synthetic peptide made to a C-terminal portion of the human Hsp90B protein (between residues 650-724) [UniProt P08238]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Antigen HSP90 beta
Quantity 0.025mL
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal