Please login to your online account to display your discounted pricing

anti-HSP90 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177682

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177682

  • / Each
 

Description & Specifications

Specifications

Antigen HSP90 alpha
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.14mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P07900
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Hsp90A protein (between residues 1-70) [UniProt P07900]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3320
Target Species Human
Target Species Murine